Bimatoprost

Lieferant: Thermo Scientific
Achtung

Synonyme: (Z)-7-((1R,2R,3R,5S)-3,5-Dihydroxy-2-((S,E)-3-hydroxy-5-phenylpent-1-en-1-yl)cyclopentyl)-N-ethylhept-5-enamide

468881000
468881000.EA 518 EUR
468881000.
Bimatoprost
Bimatoprost
Formel: C₂₅H₃₇NO₄
Molecular Weight: 415,6 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD03411999
CAS-Nummer: 155206-00-1

Bestellen

Spezifikation Testergebnisse

Appearance white to almost white powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Melting point 63°C to 67°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR