Cabozantinib S-malate 99%

Lieferant: Thermo Fisher Scientific
Achtung

468900010 468902500
468900010.EA 654 EUR
468900010. 468902500.
Cabozantinib S-malate 99%
Cabozantinib S-malate
Formel: C₃₂H₃₀FN₃O₁₀
Lagertemperatur: Kühlschrank
MDL: MFCD20923480
CAS-Nummer: 1140909-48-3

Bestellen

Spezifikation Testergebnisse

Appearance white to light beige crystalline powder
Infrared spectrum Conforms
HPLC ≥98.5 %
Melting point 166°C to 172°C
Titration with HClO4 98.5 to 101.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR