1-(2-Deoxy-2-fluoro-β-D-arabinofuranosyl)-5-iodouracil 98%
Lieferant: Thermo Scientific
Synonyme:
FIAU, Fialuridine, 2'-Deoxy-2'-fluoro-5-iodo-uridine, 1-((2R,3S,4R,5R)-3-Fluoro-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-iodopyrimidine-2,4(1H,3H)-dione, 2'-Fluoro-5-iodoarauracil, 1-[(2R,3S,4R,5R)-3-Fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodopyrimidine-2,4-dione
468971000
468971000.EA
398
EUR
468971000.
1-(2-Deoxy-2-fluoro-β-D-arabinofuranosyl)-5-iodouracil 98%
1-(2-Deoxy-2-fluoro-beta-D-arabinofuranosyl)-5-iodouracil
Formel:
C₉H₁₀FIN₂O₅ Molecular Weight: 372,09 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00866922 CAS-Nummer: 69123-98-4 Merck Index: 13,04102 |
Spezifikation Testergebnisse
Appearance | white to beige crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...