Anidulafungin 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: Ecalta, LY303366, N-[(3S,6S,9S,11R,15S,18S,20R,21R,24S,25S,26S)-6-[(1S,2S)-1,2-dihydroxy-2-(4-hydroxyphenyl)ethyl]-11,20,21,25-tetrahydroxy-3,15-bis[(1R)-1-hydroxyethyl]-26-methyl-2,5,8,14,17,23-hexaoxo-1,4,7,13,16,22-hexazatricyclo[22.3.0.09,13]heptacosan-18-yl]-4-[4-(4-pentoxyphenyl)phenyl]benzamide, Eraxis, 1-[(4R,5R)-4,5-dihydroxy-N2-[[4''-(pentyloxy)[1,1':4',1''-terphenyl]-4-yl]carbonyl]-L-ornithine]-echinocandin, N-((2R,6S,9S,11R,12R,14aS,15S,16S,20S,23S,25aS)-23-((1S,2S)-1,2-dihydroxy-2-(4-hydroxyphenyl)ethyl)-2,11,12,15-tetrahydroxy-6,20-bis((R)-1-hydroxyethyl)-16-methyl-5,8,14,19,22,25-hexaoxotetracosahydro-1H-dipyrrolo[2,1-c:2',1'-l][1,4,7,10,13,16]hexaazacyclohenicosin-9-yl)-4''-(pentyloxy)-[1,1':4',1''-terphenyl]-4-carboxamide

469081000
469081000.EA 285 EUR
469081000.
Anidulafungin 97%
Anidulafungin
Formel: C₅₈H₇₃N₇O₁₇
Molecular Weight: 1140,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00917070
CAS-Nummer: 166663-25-8

Bestellen

Spezifikation Testergebnisse

Appearance white to yellow powder
Infrared spectrum Conforms
HPLC ≥96.0 %
Water ≤8 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR