Natriumhydroxid concentrate for dilution to 0.5 N aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®

Lieferant: Avantor

Synonyme: Caustic soda

4691.EA 53.75 EUR
4691.
Natriumhydroxid concentrate for dilution to 0.5 N aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Sodium hydroxide

(1/2 equiv.=20.00 g NaOH)

Formel: NaOH
Molecular Weight: 40 g/mol
Siedepunkt: 1390
Schmelzpunkt: 323
MDL: MFCD00003548
CAS-Nummer: 1310-73-2
UN: 1823
ADR: 8 II
Merck Index: 14,08627

Bestellen

Spezifikation Testergebnisse

For Laboratory,Research,or Manufacturing Use
Normality (by titrimetry) Passes Test
(Makes 0.5N solution after dilution to 1000 mL)
Low in Carbonate


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR