Motesanib diphosphate 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
N-(3,3-Dimethylindolin-6-yl)-2-((pyridin-4-ylmethyl)amino)nicotinamide diphosphate, N-(3,3-dimethyl-1,2-dihydroindol-6-yl)-2-(pyridin-4-ylmethylamino)pyridine-3-carboxamide phosphoric acid
469181000
469181000.EA
187
EUR
469181000.
Motesanib diphosphate 98%
Motesanib diphosphate
Formel:
C₂₂H₂₉N₅O₉P₂ Molecular Weight: 569.4 g/mol Siedepunkt: 858.7°C Lagertemperatur: Raumtemperatur |
MDL:
MFCD12407403 CAS-Nummer: 857876-30-3 |
Spezifikation Testergebnisse
Appearance | white to light brown powder and or flakes |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Loss on drying | ≤5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...