N-Desmethyl clozapine 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Norclozapine

469191000
469191000.EA 401 EUR
469191000.
N-Desmethyl clozapine 98%
N-Desmethyl clozapine
Formel: C₁₇H₁₇ClN₄
Molecular Weight: 312,8 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00210189
CAS-Nummer: 6104-71-8

Bestellen

Spezifikation Testergebnisse

Appearance yellow powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR