Nutlin-3 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-(cis-4,5-Bis(4-chlorophenyl)-2-(2-isopropoxy-4-methoxyphenyl)-4,5-dihydro-1H-imidazole-1-carbonyl)piperazin-2-one, (±)-4-[4,5-Bis(4-chlorophenyl)-2-(2-isopropoxy-4-methoxy-phenyl)-4,5-dihydro-imidazole-1-carbonyl]-piperazin-2-once

469201000
469201000.EA 440 EUR
469201000.
Nutlin-3 98%
Nutlin-3
Formel: C₃₀H₃₀Cl₂N₄O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD07784509
CAS-Nummer: 548472-68-0

Bestellen

Spezifikation Testergebnisse

Appearance white to light yellow crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Loss on drying ≤5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR