Veliparib 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (R)-2-(2-Methylpyrrolidin-2-yl)-1H-benzo[d]imidazole-7-carboxamide

469261000
469261000.EA 200 EUR
469261000.
Veliparib 99%
Veliparib
Formel: C₁₃H₁₆N₄O
Molecular Weight: 244.29 g/mol
Siedepunkt: 579…580°C
Lagertemperatur: Raumtemperatur
MDL: MFCD16661059
CAS-Nummer: 912444-00-9

Bestellen

Spezifikation Testergebnisse

Appearance white to yellow powder
Infrared spectrum Conforms
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR