6,6'-Dimethyl-2,2'-bipyridin 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 6,6'-Dimethyl-2,2'-bipyridine

469310010 469310050
469310010.EA 49.6 EUR
469310010. 469310050.
6,6'-Dimethyl-2,2'-bipyridin 98%
6,6'-Dimethyl-2,2'-bipyridyl
Formel: C₁₂H₁₂N₂
Siedepunkt: 286°C
Lagertemperatur: Raumtemperatur
MDL: MFCD00059776
CAS-Nummer: 4411-80-7
EINECS: 224-566-5

Bestellen

Spezifikation Testergebnisse

Appearance white to yellow crystalline powder and or chunks
Infrared spectrum Conforms
GC ≥97.5 %
Melting point 86°C to 96°C
Water ≤1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR