6,6'-Dimethyl-2,2'-bipyridin 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
6,6'-Dimethyl-2,2'-bipyridine
469310010
469310050
469310010.EA
49.6
EUR
469310010.
469310050.
6,6'-Dimethyl-2,2'-bipyridin 98%
6,6'-Dimethyl-2,2'-bipyridyl
Formel:
C₁₂H₁₂N₂ Siedepunkt: 286°C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00059776 CAS-Nummer: 4411-80-7 EINECS: 224-566-5 |
Spezifikation Testergebnisse
Appearance | white to yellow crystalline powder and or chunks |
Infrared spectrum | Conforms |
GC | ≥97.5 % |
Melting point | 86°C to 96°C |
Water | ≤1 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...