LDK378 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
LDK-378, Ceritinib (free base), 5-Chloro-N2-(2-isopropoxy-5-methyl-4-(piperidin-4-yl)phenyl)-N4-(2-(isopropylsulphonyl)phenyl)pyrimidine-2,4-diamine
469322500
469322500.EA
197
EUR
469322500.
LDK378 98%
LDK378
Formel:
C₂₈H₃₆ClN₅O₃S Molecular Weight: 558.1 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD26142648 CAS-Nummer: 1032900-25-6 |
Spezifikation Testergebnisse
Appearance | white powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Melting point | 172°C to 177°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...