Elbasvir 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: MK-8742, Dimethyl ((2S,2'S)-((2S,2'S)-2,2'-(5,5'-((S)-6-phenyl-6H-benzo[5,6][1,3]oxazino[3,4-a]indole-3,10-diyl)bis(1H-imidazole-5,2-diyl))bis(pyrrolidine-2,1-diyl))bis(3-methyl-1-oxobutane-2,1-diyl))dicarbamate

469331000
469331000.EA 167 EUR
469331000.
Elbasvir 95%
Elbasvir
Formel: C₄₉H₅₅N₉O₇
Molecular Weight: 882.02 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28124343
CAS-Nummer: 1370468-36-2

Bestellen

Spezifikation Testergebnisse

Appearance white to light yellow powder
Infrared spectrum Conforms
HPLC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR