Glecaprevir 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
ABT-493, A 1282576, (33R,35S,91R,92R,5S,E)-5-(tert-Butyl)-N-((1R,2R)-2-(difluoromethyl)-1-(((1-methylcyclopropyl)sulphonyl)carbamoyl)cyclopropyl)-14,14-difluoro-4,7-dioxo-2,8,10-trioxa-6-aza-1(2,3)-quinoxalina-3(3,1)-pyrrolidina-9(1,2)-cyclopentanacyclotetradecaphan-12-ene-35-carboxamide
469341000
469341000.EA
622
EUR
469341000.
Glecaprevir 98%
Glecaprevir
Formel:
C₃₈H₄₆F₄N₆O₉S Molecular Weight: 838,87 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD30533436 CAS-Nummer: 1365970-03-1 |
Spezifikation Testergebnisse
Appearance | white to almost white crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...