Grazoprevir (MK-5172) 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Grazoprevir, MK-5172, (33R,35S,91R,92R,5S)-5-(tert-butyl)-N-((1R,2S)-1-((cyclopropylsulphonyl)carbamoyl)-2-vinylcyclopropyl)-17-methoxy-4,7-dioxo-2,8-dioxa-6-aza-1(2,3)-quinoxalina-3(3,1)-pyrrolidina-9(1,2)-cyclopropanacyclotetradecaphane-35-carboxamide, Zepatier
469351000
469351000.EA
252
EUR
469351000.
Grazoprevir (MK-5172) 98%
Grazoprevir (MK-5172)
Formel:
C₃₈H₅₀N₆O₉S Molecular Weight: 766,91 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD25976700 CAS-Nummer: 1350514-68-9 |
Spezifikation Testergebnisse
Appearance | white to almost white powder or crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Titration with TBAH | 97.5 to 102.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...