Pibrentasvir 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Dimethyl ((2S,2'S,3R,3'R)-((2S,2'S)-(((2R,5R)-1-(3,5-difluoro-4-(4-(4-fluorophenyl)piperidin-1-yl)phenyl)pyrrolidine-2,5-diyl)bis(5-fluoro-1H-benzo[d]imidazole-6,2-diyl))bis(pyrrolidine-2,1-diyl))bis(3-methoxy-1-oxobutane-1,2-diyl))dicarbamate, ABT-530, Dimethyl ((2S,2'S,3R,3'R)-((2S,2'S)-(((2R,5R)-1-(3,5-difluoro-4-(4-(4-fluorophenyl)piperidin-1-yl)phenyl)pyrrolidine-2,5-diyl)bis(6-fluoro-1H-benzo[d]imidazole-5,2-diyl))bis(pyrrolidine-2,1-diyl))bis(3-methoxy-1-oxobutane-1,2-diyl))dicarbamate

469371000
469371000.EA 287 EUR
469371000.
Pibrentasvir 98%
Pibrentasvir
Formel: C₅₇H₆₅F₅N₁₀O₈
Molecular Weight: 1113,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30747902
CAS-Nummer: 1353900-92-1

Bestellen

Spezifikation Testergebnisse

Appearance white to light yellow crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR