Ruxolitinib 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(R)-3-(4-(7H-Pyrrolo[2,3-d]pyrimidin-4-yl)-1H-pyrazol-1-yl)-3-cyclopentylpropanenitrile, INCB018424, Ruxolitinib
469381000
469381000.EA
297
EUR
469381000.
Ruxolitinib 98%
Ruxolitinib
Formel:
C₁₇H₁₈N₆ Molecular Weight: 306,4 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12031592 CAS-Nummer: 941678-49-5 |
Spezifikation Testergebnisse
Appearance | white to almost white powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Water | ≤2 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...