Remdesivir

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Ethylbutyl ((S)-(((2R,3S,4R,5R)-5-(4-aminopyrrolo[2,1-f][1,2,4]triazin-7-yl)-5-cyano-3,4-dihydroxytetrahydrofuran-2-yl)methoxy)(phenoxy)phosphoryl)-L-alaninate

469411000
469411000.EA 390 EUR
469411000.
Remdesivir
Remdesivir
Formel: C₂₇H₃₅N₆O₈P
Molecular Weight: 602,58 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD31657351
CAS-Nummer: 1809249-37-3

Bestellen

Spezifikation Testergebnisse

Appearance white to almost white crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR