(Z)-4-Hydroxytamoxifen 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (Z)-4-(1-(4-(2-(Dimethylamino)ethoxy)phenyl)-2-phenylbut-1-en-1-yl)phenol, (Z)-4-(1-[4-(Dimethylaminoethoxy)phenyl]-2-phenyl-1-butenyl)phenol, (Z)-4-OHT, trans-4-Hydroxytamoxifen

469501000
469501000.EA 434 EUR
469501000.
(Z)-4-Hydroxytamoxifen 99%
(Z)-4-Hydroxytamoxifen
Formel: C₂₆H₂₉NO₂
Molecular Weight: 387.51 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00941389
CAS-Nummer: 68047-06-3

Bestellen

Spezifikation Testergebnisse

Appearance white to almost white crystalline powder
Infrared spectrum Conforms
HPLC ≥98.5 %
Melting point 137°C to 143°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR