Tetrandine 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (4aS,16aS)-3,4,4a,5,16a,17,18,19-octahydro-12,21,22,26-tetramethoxy-4,17-dimethyl-16H-1,24:6,9-dietheno-11,15-metheno-2H-pyrido[2',3':17,18][1,11]dioxacycloeicosino[2,3,4-ij]isoquinoline, (1beta)-6,6',7,12-Tetramethoxy-2,2'-dimethylberbaman, (11S,31S)-16,36,37,54-Tetramethoxy-12,32-dimethyl-11,12,13,14,31,32,33,34-octahydro-2,6-dioxa-1(7,1),3(8,1)-diisoquinolina-5(1,3),7(1,4)-dibenzenacyclooctaphane, Tetrandrine, NSC 77037, d-Tetrandine

469550010
469550010.EA 215 EUR
469550010.
Tetrandine 98%
Tetrandine
Formel: C₃₈H₄₂N₂O₆
Molecular Weight: 622.8 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08689909
CAS-Nummer: 518-34-3

Bestellen

Spezifikation Testergebnisse

Appearance white to yellow to beige powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Melting point 221°C to 225°C
Water ≤5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR