Vitamin A acetat

Lieferant: Merck

Synonyme: alle-trans-Retinylacetat

46958
46958.EA 66.2 EUR
46958.
Vitamin A acetat
alle-trans-Retinylacetat
Formel: C₂₂H₃₂O₂
Molecular Weight: 328.49 g/mol
MDL: MFCD00019413
CAS-Nummer: 127-47-9
EINECS: 204-844-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=70b40cbc001975706fd5b40e35b938f2bf1e934a&vid=70b40cbc001975706fd5b40e35b938f2bf1e934a&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR