O15-Palmitoylretinol, Supelco®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

46959-U
46959-UEA 63 EUR
46959-U
O15-Palmitoylretinol, Supelco®
Vitamin A palmitate
Formel: C₃₆H₆₀O₂
MDL: MFCD00019414
CAS-Nummer: 79-81-2
EINECS: 201-228-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR