3-Carbamoylbenzoic acid 96%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
469610010
469610010.EA
218
EUR
469610010.
3-Carbamoylbenzoic acid 96%
3-Carbamoylbenzoic acid
Formel:
C₈H₇NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD04118098 CAS-Nummer: 4481-28-1 |
Spezifikation Testergebnisse
Appearance | white to beige crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥95 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...