7-Quinolinecarboxylic acid 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
469630010
469630010.EA
156
EUR
469630010.
7-Quinolinecarboxylic acid 98%
7-Quinolinecarboxylic acid
Formel:
C₁₀H₇NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00047617 CAS-Nummer: 1078-30-4 |
Spezifikation Testergebnisse
Appearance | white to beige powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Melting point | 245°C to 255°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...