3-Pyrrol-1-ylpropionic acid 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
469650010
469650010.EA
189
EUR
469650010.
3-Pyrrol-1-ylpropionic acid 97%
3-Pyrrol-1-ylpropionic acid
Formel:
C₇H₉NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD03844729 CAS-Nummer: 89059-06-3 |
Spezifikation Testergebnisse
Appearance | white to grey to brown crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Titration with NaOH | 96.0 to 104.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...