Germacrone 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(E,E)-Germacra-3,7(11),9-trien-6-one, (3E,7E)-3,7-Dimethyl-10-(propan-2-ylidene)cyclodeca-3,7-dien-1-one, trans,trans-3,7-Dimethyl-10-isopropylidene-3,7-cyclodecadien-1-one, (3E,7E)-3,7-Dimethyl-10-(propan-2-ylidene)cyclodeca-3,7-dienone
469731000
469731000.EA
322
EUR
469731000.
Germacrone 98%
Germacrone
Formel:
C₁₅H₂₂O Molecular Weight: 218,34 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00210050 CAS-Nummer: 6902-91-6 |
Spezifikation Testergebnisse
Appearance | white to almost white crystalline powder or crystals |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Melting point | 54°C to 58°C |
Water | ≤5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...