Dolutegravir sodium salt 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Sodium (4R,12aS)-9-((2,4-difluorobenzyl)carbamoyl)-4-methyl-6,8-dioxo-3,4,6,8,12,12a-hexahydro-2H-pyrido[1',2':4,5]pyrazino[2,1-b][1,3]oxazin-7-olate, GSK1349572, Dolutegravir sodium

469770010 469772500
469770010.EA 198 EUR
469770010. 469772500.
Dolutegravir sodium salt 99%
Dolutegravir sodium salt
Formel: C₂₀H₁₈F₂N₃NaO₅
Lagertemperatur: Kühlschrank
MDL: MFCD28405599
CAS-Nummer: 1051375-19-9
UN: 3077
ADR: 9,III

Bestellen

Spezifikation Testergebnisse

Appearance white to light yellow powder or crystals
Infrared spectrum Conforms
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR