Ibudilast 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1-(2-Isopropylpyrazolo[1,5-a]pyridin-3-yl)-2-methylpropan-1-one, 3-Isobutyryl-2-isopropylpyrazolo[1,5-a]pyridine

469782500
469782500.EA 92.8 EUR
469782500.
Ibudilast 98%
Ibudilast
Formel: C₁₄H₁₈N₂O
Molecular Weight: 230.31 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00864808
CAS-Nummer: 50847-11-5
Merck Index: 13,04904

Bestellen

Spezifikation Testergebnisse

Appearance white to almost white crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Melting point 54°C to 58°C
Water ≤0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR