Lapatinib ditosylate monohydrate 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
N-(3-Chloro-4-((3-fluorobenzyl)oxy)phenyl)-6-(5-(((2-(methylsulfonyl)ethyl)amino)methyl)furan-2-yl)quinazolin-4-amine bis(4-methylbenzenesulfonate) hydrate, GW2016 ditosylate monohydrate, Lapatinib ditosylate hydrate, Lapatinib tosilate hydrate, Lapatinib tosilate, GW572016 ditosylate monohydrate
469800010
469800010.EA
83.2
EUR
469800010.
Lapatinib ditosylate monohydrate 98%
Lapatinib ditosylate monohydrate
Formel:
C₄₃H₄₄ClFN₄O₁₁S₃ Molecular Weight: 943,48 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD18904381 CAS-Nummer: 388082-78-8 |
Spezifikation Testergebnisse
Appearance | yellow crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Water | 1.0 to 3.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...