Methyl mercaptan sodium salt 90+%, technical

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Sodium methanethiolate, Sodium thiomethoxide, Sodium methylsulphanide, Methanethiol sodium salt

469890010 469890050 469890250 469891000
469890010.EA 32.7 EUR
469890010. 469890050. 469890250. 469891000.
Methyl mercaptan sodium salt 90+%, technical
Methyl mercaptan sodium salt
Formel: CH₃NaS
Molecular Weight: 70.08 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00174316
CAS-Nummer: 5188-07-8
EINECS: 225-969-9
UN: 2921
ADR: 8 I

Bestellen

Spezifikation Testergebnisse

Appearance white to light yellow to light brown powder or granular powder
Titration Iodimetric ≥90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR