Methyl mercaptan sodium salt 90+%, technical
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
469890010
469890050
469890250
469891000
469890010.EA
32.7
EUR
469890010.
469890050.
469890250.
469891000.
Methyl mercaptan sodium salt 90+%, technical
Methyl mercaptan sodium salt
Formel:
CH₃NaS Lagertemperatur: Raumtemperatur |
MDL:
MFCD00174316 CAS-Nummer: 5188-07-8 EINECS: 225-969-9 UN: 2921 ADR: 8,I |
Spezifikation Testergebnisse
Appearance | white to light yellow to light brown powder or granular powder |
Titration Iodimetric | ≥90.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...