Rhodium(IV)oxid (75.2 - 77.4% Rh)

Lieferant: Thermo Fisher Scientific

47002.MD 47002.03
47002.MDEA 0 EUR
47002.MD 47002.03
Rhodium(IV)oxid (75.2 - 77.4% Rh)
Rhodium(IV) oxide
Formel: O₂Rh
Dichte: 11,4 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
CAS-Nummer: 12137-27-8
EINECS: 235-237-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR