(R)-Roscovitine 98%

Lieferant: Thermo Scientific

Synonyme: 2-(R)-(1-Ethyl-2-hydroxyethylamino)-6-benzylamino-9-isopropylpurine, Roscovitine, (R)-2-((6-(Benzylamino)-9-isopropyl-9H-purin-2-yl)amino)butan-1-ol

470030500
470030500.EA 180 EUR
470030500.
(R)-Roscovitine 98%
(R)-Roscovitine
Formel: C₁₉H₂₆N₆O
Molecular Weight: 354,45 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD02266401
CAS-Nummer: 186692-46-6

Bestellen

Spezifikation Testergebnisse

Appearance white to light yellow powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Melting point 104°C to 108°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR