Salvianolic acid C 95%
Lieferant: Thermo Scientific
Synonyme:
(R,E)-3-(3,4-Dihydroxyphenyl)-2-((3-(2-(3,4-dihydroxyphenyl)-7-hydroxybenzofuran-4-yl)acryloyl)oxy)propanoic acid
470040100
470040100.EA
287
EUR
470040100.
Salvianolic acid C 95%
Salvianolic acid C
Formel:
C₂₆H₂₀O₁₀ Molecular Weight: 492,43 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD16660675 CAS-Nummer: 115841-09-3 |
Spezifikation Testergebnisse
Appearance | white to yellow powder |
Infrared spectrum | Conforms |
HPLC | ≥94 % |
Water | ≤15 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...