Shikonin 99%

Lieferant: Thermo Scientific

Synonyme: (R)-5,8-Dihydroxy-2-(1-hydroxy-4-methylpent-3-en-1-yl)naphthalene-1,4-dione, Tokyo Violet, 5,8-Dihydroxy-2-[(1R)-1-hydroxy-4-methyl-3-penten-1-yl]-1,4-naphthalenedione, C.I. 75535

470120500
470120500.EA 179 EUR
470120500.
Shikonin 99%
Shikonin
Formel: C₁₆H₁₆O₅
Molecular Weight: 288,3 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00016668
CAS-Nummer: 517-89-5

Bestellen

Spezifikation Testergebnisse

Appearance red to brown powder
Infrared spectrum Conforms
HPLC ≥98.5 %
Melting point 139°C to 142°C
Water ≤1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR