Levosimendan 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
OR-1259, 2-[2-[4-[(4R)-1,4,5,6-Tetrahydro-4-methyl-6-oxo-3-pyridazinyl]phenyl]hydrazinylidene]propanedinitrile, (R)-N-(4-(4-Methyl-6-oxo-1,4,5,6-tetrahydropyridazin-3-yl)phenyl)carbonohydrazonoyl dicyanide, (R)-Simendan
470190010
470190010.EA
272
EUR
470190010.
Levosimendan 98%
Levosimendan
Formel:
C₁₄H₁₂N₆O Molecular Weight: 280,29 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00867135 CAS-Nummer: 141505-33-1 Merck Index: 13,05491 |
Spezifikation Testergebnisse
Appearance | yellow to light brown powder or crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Optical rotation | -500° to -650° (0.5% in THF/MeOH 1/1) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...