Dansylhydrazid 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
5-(Dimethylamino)naphthalinsulfonsäurehydrazid, 5-(Dimethylamino)naphthalinsulfonohydrazid, 5-(Dimethylamino)naphthalin-1-sulfonohydrazid, Dansylhydrazin
470230010
470230010.EA
58.4
EUR
470230010.
Dansylhydrazid 97%
Dansylhydrazid
Formel:
C₁₂H₁₅N₃O₂S Molecular Weight: 265,34 g/mol Siedepunkt: 450,8°C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003986 CAS-Nummer: 33008-06-9 EINECS: 251-337-7 |
Spezifikation Testergebnisse
Appearance | white to yellow to green crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Melting point | 126°C to 130°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...