Mangan(III)acetat Dihydrat ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

47050.04 47050.09 47050.18
47050.04EA 112 EUR
47050.04 47050.09 47050.18
Mangan(III)acetat Dihydrat ≥97%
Mangan(III)acetat Dihydrat
Formel: (H₃CCOO)₂Mn·2H₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00150022
CAS-Nummer: 19513-05-4
EINECS: 213-602-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR