Cobalt bis(trifluoromethylsulphonyl)imide

Lieferant: Thermo Fisher Scientific
Gefahr

47053.03 47053.06
47053.03EA 126 EUR
47053.03 47053.06
Cobalt bis(trifluoromethylsulphonyl)imide
Cobalt bis(trifluoromethylsulphonyl)imide

CAS No.: 207861-61-8 Molecular Formula: C4CoF12N2O8S4 Formula Weight: 619.23 Physical Form: Solid Sensitivity: Hygroscopic Appearance: Pale purple to purple Melting Point: 200° UN No.: UN3261 MDL No.: MFCD23380169

Formel: C₄CoF₁₂N₂O₈S₄
Schmelzpunkt: >200 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD23380169
CAS-Nummer: 207861-61-8
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR