N-Methyl-N-(1-methylethyl)glycine 90%

Lieferant: Thermo Fisher Scientific

Synonyme: 2-(Isopropyl(methyl)amino)acetic acid

470622500
470622500.EA 116 EUR
470622500.
N-Methyl-N-(1-methylethyl)glycine 90%
N-Methyl-N-(1-methylethyl)glycine
Formel: C₆H₁₃NO₂
MDL: MFCD09864552
CAS-Nummer: 108957-96-6

Bestellen

Spezifikation Testergebnisse

Appearance white to brown solid
Infrared spectrum Conforms
Titration Tertiary base 90.0 to 110.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR