Y-27632·dihydrochloride 99%

Lieferant: Thermo Fisher Scientific

Synonyme: Y-27632 2HCl, (R)-(+)-trans-4-(1-Aminoethyl)-N-(4-Pyridyl)cyclohexanecarboxamide dihydrochloride, Rock inhibitor y-27632, trans-4-((R)-1-Aminoethyl)-N-(pyridin-4-yl)cyclohexanecarboxamide dihydrochloride

470701000
470701000.EA 444 EUR
470701000.
Y-27632·dihydrochloride 99%
Y-27632 dihydrochloride
Formel: C₁₄H₂₁N₃O·2HCl
Siedepunkt: 258°C (760mmHg)
Lagertemperatur: Kühlschrank
MDL: MFCD03490488
CAS-Nummer: 129830-38-2 | 146986-50-7 (free base)

Bestellen

Spezifikation Testergebnisse

Appearance white to beige powder
Infrared spectrum Conforms
HPLC ≥98.5 %
Water ≤8 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR