Naratriptan hydrochloride 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: N-Methyl-3-(1-methyl-4-piperidinyl)-1H-indole-5-ethanesulphonamide hydrochloride

470752500
470752500.EA 134 EUR
470752500.
Naratriptan hydrochloride 95%
Naratriptan hydrochloride
Formel: C₁₇H₂₅N₃O₂S·HCl
Molecular Weight: 371,92 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08141821
CAS-Nummer: 143388-64-1

Bestellen

Spezifikation Testergebnisse

Appearance white to beige powder
Infrared spectrum Conforms
HPLC ≥94 %
Melting point 234°C to 246°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR