2-(Hydroxymethyl)-1,3-propandiol 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Tris(hydroxymethyl)methane, 2-(Hydroxymethyl)propane-1,3-diol

470770010
470770010.EA 112 EUR
470770010.
2-(Hydroxymethyl)-1,3-propandiol 98%
2-(Hydroxymethyl)-1,3-propanediol
Formel: C₄H₁₀O₃
Molecular Weight: 106,12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00192217
CAS-Nummer: 4704-94-3
EINECS: 225-187-8

Bestellen

Spezifikation Testergebnisse

Appearance white powder or crystals or chunks
Infrared spectrum Conforms
GC ≥97.5 %
Melting point 63°C to 68°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR