Nimustinhydrochlorid 95%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Nimustine HCl, 3-((4-Amino-2-methylpyrimidin-5-yl)methyl)-1-(2-chloroethyl)-1-nitrosourea hydrochloride, ACNU, 1-[(4-Amino-2-methyl-5-pyrimidinyl)methyl]-3-(2-chloroethyl)-3-nitrosourea Hydrochloride
470860010
470860010.EA
121
EUR
470860010.
Nimustinhydrochlorid 95%
Nimustinhydrochlorid
Formel:
C₉H₁₃ClN₆O₂ Molecular Weight: 309,15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01676942 CAS-Nummer: 55661-38-6 UN: 2811 ADR: 6.1,III Merck Index: 13,06581 |
Spezifikation Testergebnisse
Appearance | white to light yellow powder or crystals |
Infrared spectrum | Conforms |
HPLC | ≥94 % |
Titration with HClO4 | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...