Di-tert-butyloxalate 96%

Lieferant: Thermo Fisher Scientific
Achtung

470960050 470960250
470960050.EA 64.8 EUR
470960050. 470960250.
Di-tert-butyloxalate 96%
Di-tert-butyloxalate
Formel: C₁₀H₁₈O₄
Siedepunkt: 229,3°C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008806
CAS-Nummer: 691-64-5
EINECS: 211-723-8

Bestellen

Spezifikation Testergebnisse

Appearance colorless to almost white powder or crystals
Infrared spectrum Conforms
GC ≥95 %
Melting point 69°C to 72°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR