Laborkühlschränke, ATEX
Lieferant: Liebherr
LFKEX3910 LFKUEX1610
471-0644EA 0 EUR
471-0644 471-0647
Laborkühlschränke, ATEX
Kühlschränke
Interior satisfies the requirement, for storage of volatile solvents, of the ATEX Directive 94/9 EC, rated as II 3G Ex nA II T6.
  • Digital control and display
  • Audible and visual alarm for temperature with interface for remote alarm
  • Audible and visual alarm for open door status (on the equipment - not recorded)
  • Recording of temperature alarms
  • RS485 interface for PC control and traceability
Interior has housing for probe diameter 30 mm for the control / qualification.
Predrilled access port for PT100 (not supplied) probe diameter 8 mm for the independent temperature monitoring of the interior.
Temperature control: +5 °C ±3 °C .
Dynamic cooling.
Auto defrost.
Temperature control: +5 °C ±3 °C