Laboratory refrigerator, LABEX® 105
Lieferant: KIRSCH PHILIPP
10298
471-2528EA 3400 EUR
471-2528
Laboratory refrigerator, LABEX® 105
Kühlschränke
This laboratory refrigerator is made of from galvanised sheet steel (rustproof), with white, anti-scratch powder coating.
  • Adjustable feet, automatic defrosting
  • Lockable door with easy-to-replace plastic magnetic seal frame
  • Control and display sensors protected by safety barriers
  • Antifreeze against sub-zero temperatures
  • Ventilation-enforced refrigerating machine
  • Power failure alarm
Key switch protected control panel against tampering. Membrane keyboard with digital temperature display and minimum/maximum temperature memory.
Interior consists of one drawer with stops and two plasticcoated shelves.