External housing of laboratory refrigerator are made of galvanised sheet steel (rustproof), with white, anti-scratch powder coating. Interior are made of smooth aluminium with colourless protective coating.
- Low-noise compressor
- Automatic defrosting
- Re-circulating air cooling
- Removable condensation container
- Antifreeze against sub-zero temperatures
- Ventilation-enforced refrigerating machine
Self-closing door with easy-to-replace plastic magnetic seal frame, lockable. Forced-air cooling (axial fan is proteced), switches off automatically when the door opens, ensures a uniform temperature and minimises temperature deviations. RS485 interface for connecting to PC supported temperature documentation.
Standards: Explosion proof acc. to ATEX 95.