Laboratory refrigerator, LABEX®-468
Lieferant: KIRSCH PHILIPP
10569
471-2532EA 5670 EUR
471-2532
Laboratory refrigerator, LABEX®-468
Kühlschränke
External housing of laboratory refrigerator are made of galvanised sheet steel (rustproof), with white, anti-scratch powder coating. Interior are made of smooth aluminium with colourless protective coating.
  • Low-noise compressor
  • Automatic defrosting
  • Re-circulating air cooling
  • Removable condensation container
  • Antifreeze against sub-zero temperatures
  • Ventilation-enforced refrigerating machine
Self-closing door with easy-to-replace plastic magnetic seal frame, lockable. Forced-air cooling (axial fan is proteced), switches off automatically when the door opens, ensures a uniform temperature and minimises temperature deviations. RS485 interface for connecting to PC supported temperature documentation.
Standards: Explosion proof acc. to ATEX 95.