Laboratory refrigerator, SUPER-720-CHROMAT
Lieferant: KIRSCH PHILIPP
10480
471-2550EA 8360 EUR
471-2550
Laboratory refrigerator, SUPER-720-CHROMAT
Kühlschränke
Outer panels of laboratory refrigerator are made of galvanised sheet steel (protection against sub surface rusting), white powder coated. Interior made of smooth aluminium sheet with clear protective coating.
  • Recirculating air-cooling
  • GSM-module
  • RS 485 Interface
  • Safety control device prevents freezing
  • Compressor, static ventilated, hermetically enclosed
  • Automatic rapid defrosting with melt water evaporation
Key switch protected control panel with digital temperature display, minimum/maximum temperature memory, optical and acoustic warning signal in case of temperature deviation and door ajar alarm with potenial free contact for remote monitoring. Switch for air-humidity selection, defrost control light and red warning light. Temperature gear by electronic thermostat.