Indium(III)iodid, ultra trocken ≥99,999% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Unter Argon in Ampullen verpackt
Purity: 99.999%
CAS No.: 13510-35-5
Molecular Formula: InI3
Formula Weight: 495.53
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 4.69
Sensitivity: Light Sensitive; Hygroscopic
Appearance: Yellow to brown
Melting Point: 210°
UN No.: UN3260
MDL No.: MFCD00016151
Formel:
I₃In Schmelzpunkt: 210 °C Dichte: 4,69 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016151 CAS-Nummer: 13510-35-5 EINECS: 236-839-6 UN: 3260 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...